siRNA Details
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108051.1 | NP_001101521.1 | 10 | 1103 | ||
XM_006240471.2 | XP_006240533.1 | 13 | 4570 | |||
XM_006240472.2 | XP_006240534.1 | 13 | 4513 | |||
GenBank mRNA | BC168870.1 | - | 12 | 1383 | 7104573 |
