siRNA Details
Gene Symbol: Scyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191780.1 | NP_001178709.1 | 4 | 641 | ||
XM_006241241.2 | XP_006241303.1 | 4 | 678 | |||
XM_006241242.2 | XP_006241304.1 | 4 | 678 |
