siRNA Details
Gene Symbol: Acss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108091.1 | NP_001101561.1 | 8 | 1235 | ||
XM_006241302.2 | XP_006241364.1 | 8 | 1233 | |||
XM_006241303.2 | XP_006241365.1 | 8 | 1922 | |||
XM_008765327.1 | XP_008763549.1 | 8 | 973 |
