siRNA Details
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750319.3 | XP_003750367.1 | 23, 24 | 2971 | ||
XM_006241324.2 | XP_006241386.1 | 22, 23 | 3967 | |||
XM_006241325.1 | XP_006241387.1 | 21, 22 | 2538 | |||
XM_006241326.2 | XP_006241388.1 | 21, 22 | 3930 | |||
XM_008765365.1 | XP_008763587.1 | 23, 24 | 2690 | |||
XM_008765367.1 | XP_008763589.1 | 22, 23 | 9395 | |||
GenBank mRNA | BC168899.1 | - | 4, 5 | 842 | 7455687 |
