siRNA Details
Gene Symbol: Osbpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750319.3 | XP_003750367.1 | 24 | 3036 | ||
XM_006241324.2 | XP_006241386.1 | 23 | 4032 | |||
XM_006241325.1 | XP_006241387.1 | 22 | 2603 | |||
XM_006241326.2 | XP_006241388.1 | 22 | 3995 | |||
XM_008765365.1 | XP_008763587.1 | 24 | 2755 | |||
XM_008765367.1 | XP_008763589.1 | 23 | 9460 | |||
GenBank mRNA | BC168899.1 | - | 5 | 907 | 7455687 |
