siRNA Details
Gene Symbol: Best3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191783.1 | NP_001178712.1 | 9 | 1259 | ||
XM_008765394.1 | XP_008763616.1 | 11 | 2403 | |||
XM_008765395.1 | XP_008763617.1 | 9 | 2140 | |||
GenBank mRNA | FQ222375.1 | - | 11 | 1754 |
