siRNA Details
Gene Symbol: U2surp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729954.4 | XP_002730000.1 | 16, 17 | 1728 | ||
XM_006243554.1 | XP_006243616.1 | 17, 18 | 1873 | |||
XM_236501.7 | XP_236501.3 | 17, 18 | 1914 | |||
GenBank mRNA | FQ226547.1 | - | 17, 18 | 1882 |
