siRNA Details
Gene Symbol: Nme9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766584.1 | XP_008764806.1 | 5 | 275 | ||
XM_008766585.1 | XP_008764807.1 | 5 | 274 | |||
XM_008766586.1 | XP_008764808.1 | 5 | 275 | |||
XM_008766587.1 | XP_008764809.1 | 5 | 274 |
