siRNA Details
Gene Symbol: Dzip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014095.1 | NP_001014117.1 | 2 | 334 | ||
XM_006243626.2 | XP_006243688.1 | 3 | 538 | |||
XM_006243627.2 | XP_006243689.1 | 3 | 689 | |||
XM_008766526.1 | XP_008764748.1 | 2 | 1302 | |||
XM_008766527.1 | XP_008764749.1 | 2 | 1302 | |||
GenBank mRNA | BC083786.1 | - | 2 | 334 | 7097259 |
