siRNA Details
Gene Symbol: Topbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243684.1 | XP_006243746.1 | 13 | 2279 | ||
XM_008766589.1 | XP_008764811.1 | 13 | 2079 | |||
XM_236578.7 | XP_236578.6 | 13 | 2279 |
