siRNA Details
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244025.2 | XP_006244087.1 | 6 | 1010 | ||
XM_008766657.1 | XP_008764879.1 | 6 | 1067 | |||
XM_243980.7 | XP_243980.4 | 6 | 2162 |
