siRNA Details
Gene Symbol: Azi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025705.1 | NP_001020876.1 | 8 | 1427 | ||
XM_006244032.2 | XP_006244094.1 | 9 | 2298 | |||
GenBank mRNA | BC098680.1 | - | 8 | 1427 | 7384893 | |
FQ221679.1 | - | 9 | 1547 |
