siRNA Details
Gene Symbol: Xylb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033704.1 | NP_001028876.1 | 15 | 1244 | ||
XM_006244093.1 | XP_006244155.1 | 16 | 1616 | |||
XM_006244094.1 | XP_006244156.1 | 16 | 1616 | |||
XM_006244095.1 | XP_006244157.1 | 16 | 1616 | |||
XM_006244096.1 | XP_006244158.1 | 14 | 1256 | |||
GenBank mRNA | BC101852.1 | - | 15 | 1244 | 7930657 |
