siRNA Details
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108230.1 | NP_001101700.1 | 15 | 1884 | ||
XM_006245396.2 | XP_006245458.2 | 19 | 6289 | |||
XM_006245397.2 | XP_006245459.2 | 19 | 6253 | |||
XM_006245398.2 | XP_006245460.2 | 18 | 6174 | |||
XM_006245400.2 | XP_006245462.2 | 17 | 6141 | |||
XM_006245401.2 | XP_006245463.2 | 18 | 6130 | |||
XM_006245402.2 | XP_006245464.2 | 18 | 6073 | |||
XM_006245403.2 | XP_006245465.2 | 16 | 5982 | |||
GenBank mRNA | BC158781.1 | - | 2 | 257 |
