siRNA Details
Gene Symbol: Smchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_244261.8 | XP_244261.6 | 3 | 704 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Smchd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_244261.8 | XP_244261.6 | 3 | 704 |