siRNA Details
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108236.1 | NP_001101706.1 | 11 | 1675 | ||
XM_006245662.2 | XP_006245724.1 | 11 | 3979 | |||
XM_006245663.2 | XP_006245725.1 | 11 | 4996 | |||
XM_006245664.2 | XP_006245726.1 | 11 | 2224 | |||
XM_006245665.2 | XP_006245727.1 | 11 | 3981 | |||
XM_006245666.2 | XP_006245728.1 | 11 | 1713 |
