siRNA Details
Gene Symbol: Brcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127300.1 | NP_001120772.1 | 1 | 57 | ||
GenBank mRNA | BC166833.1 | - | 1 | 57 | 9021884 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Brcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127300.1 | NP_001120772.1 | 1 | 57 | ||
GenBank mRNA | BC166833.1 | - | 1 | 57 | 9021884 |