siRNA Details
Gene Symbol: Uprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257154.2 | XP_006257216.1 | 1 | 462 | ||
XM_228538.8 | XP_228538.3 | 2 | 414 |

More Information
Other Species Targeted

Gene Ontology Categories:
