siRNA Details
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191725.2 | NP_001178654.1 | 6, 7 | 734 | ||
XM_006254380.2 | XP_006254442.1 | 7, 8 | 826 | |||
XM_006254381.2 | XP_006254443.1 | 7, 8 | 798 |
