siRNA Details
Gene Symbol: Crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022501.1 | NP_071946.1 | 5, 6 | 555 | ||
XM_008764943.1 | XP_008763165.1 | 5, 6 | 819 | |||
GenBank mRNA | BC061774.1 | - | 5, 6 | 524 | 5621903 | |
D17512.1 | - | 5, 6 | 555 | |||
FQ226314.1 | - | 5, 6 | 563 | |||
FQ227529.1 | - | 5, 6 | 565 | |||
FQ230331.1 | - | 5, 6 | 565 | |||
FQ233584.1 | - | 5, 6 | 569 |
