siRNA Details
Gene Symbol: Crip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022501.1 | NP_071946.1 | 7 | 634 | ||
XM_008764943.1 | XP_008763165.1 | 7 | 898 | |||
GenBank mRNA | BC061774.1 | - | 7 | 603 | 5621903 | |
D17512.1 | - | 7 | 634 | |||
FQ226314.1 | - | 7 | 642 | |||
FQ227529.1 | - | 7 | 644 | |||
FQ230331.1 | - | 7 | 644 | |||
FQ233584.1 | - | 7 | 648 |
