siRNA Details
Gene Symbol: Ptprk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029902.2 | NP_001025073.2 | 24 | 3594 | ||
XM_008758616.1 | XP_008756838.1 | 26 | 3845 | |||
XM_008758617.1 | XP_008756839.1 | 25 | 3826 | |||
XM_008758618.1 | XP_008756840.1 | 25 | 3808 | |||
GenBank mRNA | AB288088.1 | - | 11 | 1329 | ||
AB297790.1 | - | 24 | 3594 |
