siRNA Details
Gene Symbol: Figf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031761.1 | NP_113949.1 | 6 | 1156 | ||
XM_006256863.2 | XP_006256925.1 | 7 | 1303 | |||
XM_006256864.2 | XP_006256926.1 | 6 | 1633 | |||
GenBank mRNA | AF014827.1 | - | 6 | 1156 | ||
AY032728.1 | - | 6 | 931 |
