siRNA Details
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108291.1 | NP_001101761.1 | 11 | 1476 | ||
XM_006247155.2 | XP_006247217.1 | 11 | 1468 | |||
XM_008768078.1 | XP_008766300.1 | 10 | 1572 | |||
GenBank mRNA | FQ212347.1 | - | 10 | 1552 |
