siRNA Details
Gene Symbol: Epn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024791.1 | NP_001019962.1 | 2 | 234 | ||
XM_006247201.2 | XP_006247263.1 | 2 | 1337 | |||
XM_006247202.2 | XP_006247264.1 | 2 | 1302 | |||
XM_006247203.2 | XP_006247265.1 | 2 | 1337 | |||
GenBank mRNA | BC097500.1 | - | 2 | 234 | 7462750 |
