siRNA Details
Gene Symbol: Mfsd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108308.1 | NP_001101778.1 | 4 | 550 | ||
XM_006247809.2 | XP_006247871.1 | 5 | 898 | |||
XM_006247810.2 | XP_006247872.1 | 5 | 499 | |||
XM_006247811.2 | XP_006247873.1 | 3 | 231 |
