siRNA Details
Gene Symbol: Mfsd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108308.1 | NP_001101778.1 | 7 | 818 | ||
XM_006247809.2 | XP_006247871.1 | 8 | 1166 | |||
XM_006247810.2 | XP_006247872.1 | 8 | 767 | |||
XM_006247811.2 | XP_006247873.1 | 6 | 499 |
