siRNA Details
Gene Symbol: RGD1563888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108312.2 | NP_001101782.1 | 452 | |||
XM_008768531.1 | XP_008766753.1 | 698 | ||||
XM_008768532.1 | XP_008766754.1 | 733 | ||||
GenBank mRNA | FQ227934.1 | - | 710 |
