siRNA Details
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047899.2 | NP_001041364.1 | 5 | 484 | ||
XM_008768736.1 | XP_008766958.1 | 7 | 690 | |||
XM_008768737.1 | XP_008766959.1 | 7 | 742 | |||
GenBank mRNA | AY310146.1 | - | 5 | 524 | ||
AY724512.1 | - | 8 | 968 |
