siRNA Details
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007147.1 | NP_001007148.1 | 15, 16 | 1851 | ||
XM_006248941.2 | XP_006249003.1 | 19, 20 | 2385 | |||
XM_006248942.2 | XP_006249004.1 | 19, 20 | 2382 | |||
XM_006248943.1 | XP_006249005.1 | 20, 21 | 2465 | |||
XM_006248945.1 | XP_006249007.1 | 19, 20 | 2355 | |||
XM_006248946.1 | XP_006249008.1 | 20, 21 | 2437 | |||
XM_006248947.2 | XP_006249009.1 | 19, 20 | 3591 | |||
XM_006248948.2 | XP_006249010.1 | 18, 19 | 2274 | |||
XM_006248949.2 | XP_006249011.1 | 18, 19 | 2187 | |||
XM_006248951.2 | XP_006249013.1 | 17, 18 | 2076 | |||
XM_006248952.2 | XP_006249014.1 | 16, 17 | 2010 | |||
XM_006248953.2 | XP_006249015.1 | 15, 16 | 1911 | |||
XM_008768982.1 | XP_008767204.1 | 21, 22 | 2615 | |||
XM_008768983.1 | XP_008767205.1 | 19, 20 | 2513 | |||
XM_008768984.1 | XP_008767206.1 | 18, 19 | 2233 | |||
GenBank mRNA | BC085844.1 | - | 15, 16 | 1851 | 7135918 |
