siRNA Details
Gene Symbol: Rasal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108335.1 | NP_001101805.1 | 20, 21 | 2463 | ||
XM_006249395.2 | XP_006249457.1 | 19, 20 | 3660 | |||
XM_006249396.2 | XP_006249458.1 | 19, 20 | 3664 |
