siRNA Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249782.2 | XP_006249844.1 | 22 | 3314 | ||
XM_006249783.2 | XP_006249845.1 | 22 | 3198 | |||
XM_006249784.2 | XP_006249846.1 | 22 | 3312 | |||
XM_006249785.2 | XP_006249847.1 | 21 | 4263 | |||
XM_008769479.1 | XP_008767701.1 | 22 | 3309 | |||
XM_008769480.1 | XP_008767702.1 | 19 | 2231 | |||
XM_008769481.1 | XP_008767703.1 | 19 | 2160 | |||
GenBank mRNA | FQ212420.1 | - | 15 | 1631 |
