siRNA Details
Gene Symbol: Tbc1d14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012152.2 | NP_001012152.2 | 7 | 677 | ||
NM_001034021.2 | NP_001029193.2 | 9 | 1539 | |||
NM_001113365.1 | NP_001106836.1 | 10 | 1766 | |||
XM_006251133.2 | XP_006251195.1 | 10 | 1772 | |||
XM_006251134.2 | XP_006251196.1 | 9 | 1464 | |||
XM_008770323.1 | XP_008768545.1 | 10 | 1953 | |||
XM_008770324.1 | XP_008768546.1 | 8 | 3461 | |||
XM_008770325.1 | XP_008768547.1 | 7 | 614 | |||
XM_008770326.1 | XP_008768548.1 | 7 | 657 | |||
GenBank mRNA | AB108668.1 | - | 7 | 614 | ||
AB117752.1 | - | 10 | 1766 | |||
BC099760.1 | - | 9 | 1512 | 7930485 |
