siRNA Details
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108365.1 | NP_001101835.1 | 3 | 488 | ||
XM_006251340.2 | XP_006251402.1 | 2 | 523 | |||
XM_008770332.1 | XP_008768554.1 | 1 | 849 | |||
XM_008770333.1 | XP_008768555.1 | 1 | 848 |
