siRNA Details
Gene Symbol: RGD1310110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108374.1 | NP_001101844.1 | 2 | 718 | ||
GenBank mRNA | FQ212344.1 | - | 2 | 720 |
Gene Symbol: LOC689945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770651.1 | XP_008768873.1 | 3 | 940 |
