siRNA Details
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108384.1 | NP_001101854.1 | 3 | 397 | ||
XM_006252275.2 | XP_006252337.1 | 3 | 439 | |||
XM_006252276.2 | XP_006252338.1 | 3 | 842 | |||
XM_006252277.2 | XP_006252339.1 | 3 | 439 |
