siRNA Details
Gene Symbol: Dgkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252363.2 | XP_006252425.1 | 16 | 2077 | ||
XM_006252364.2 | XP_006252426.1 | 16 | 4394 | |||
XM_008770918.1 | XP_008769140.1 | 18 | 3066 | |||
XM_008770919.1 | XP_008769141.1 | 18 | 3065 | |||
XM_008770920.1 | XP_008769142.1 | 16 | 2142 |
