siRNA Details
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127494.1 | NP_001120966.1 | 387 | |||
GenBank mRNA | EU371958.1 | - | 387 |
Gene Symbol: LOC501266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759826.1 | XP_008758048.1 | 1 | 699 |
