siRNA Details
Gene Symbol: Fgd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108409.1 | NP_001101879.1 | 9 | 1395 | ||
XM_008771543.1 | XP_008769765.1 | 9 | 1946 | |||
XM_008771544.1 | XP_008769766.1 | 9 | 2554 | |||
XM_008771545.1 | XP_008769767.1 | 9 | 1648 | |||
XM_008771546.1 | XP_008769768.1 | 9 | 1617 |

More Information
Other Species Targeted

Gene Ontology Categories:
