siRNA Details
Gene Symbol: Svil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108416.2 | NP_001101886.2 | 36, 37 | 7022 | ||
XM_006254030.2 | XP_006254092.1 | 40, 41 | 7154 | |||
XM_006254031.2 | XP_006254093.1 | 38, 39 | 8892 | |||
XM_008771737.1 | XP_008769959.1 | 40, 41 | 9244 | |||
XM_008771738.1 | XP_008769960.1 | 40, 41 | 6814 | |||
XM_008771739.1 | XP_008769961.1 | 39, 40 | 7045 | |||
XM_008771740.1 | XP_008769962.1 | 39, 40 | 6825 | |||
XM_008771741.1 | XP_008769963.1 | 40, 41 | 7151 | |||
XM_008771742.1 | XP_008769964.1 | 39, 40 | 7076 | |||
XM_008771743.1 | XP_008769965.1 | 39, 40 | 7057 | |||
XM_008771744.1 | XP_008769966.1 | 38, 39 | 7027 | |||
XM_008771745.1 | XP_008769967.1 | 38, 39 | 7024 | |||
XM_008771746.1 | XP_008769968.1 | 39, 40 | 6988 | |||
XM_008771747.1 | XP_008769969.1 | 38, 39 | 6944 | |||
XM_008771748.1 | XP_008769970.1 | 38, 39 | 6132 | |||
XM_008771749.1 | XP_008769971.1 | 37, 38 | 6048 | |||
XM_008771750.1 | XP_008769972.1 | 34, 35 | 5825 |
