siRNA Details
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013191.1 | NP_001013209.1 | 3 | 469 | ||
XM_006255502.2 | XP_006255564.1 | 3 | 463 | |||
GenBank mRNA | BC081946.1 | - | 3 | 469 | 7121452 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_022309, NM_001161457, NM_001161458, BC006763, L03279, and others.
Gene Ontology Categories:
