siRNA Details
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108453.1 | NP_001101923.1 | 12 | 1825 | ||
XM_006255770.2 | XP_006255832.1 | 11 | 2719 | |||
XM_006255772.2 | XP_006255834.1 | 12 | 1980 | |||
XM_008772629.1 | XP_008770851.1 | 11 | 3690 | |||
XM_008772630.1 | XP_008770852.1 | 10 | 3613 | |||
XM_008772631.1 | XP_008770853.1 | 11 | 2175 |
