siRNA Details
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108455.1 | NP_001101925.1 | 34 | 3379 | ||
XM_008772633.1 | XP_008770855.1 | 35 | 3503 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108455.1 | NP_001101925.1 | 34 | 3379 | ||
XM_008772633.1 | XP_008770855.1 | 35 | 3503 |