siRNA Details
Gene Symbol: Ube2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108471.1 | NP_001101941.1 | 2 | 763 | ||
GenBank mRNA | FQ209516.1 | - | 3 | 356 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ube2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108471.1 | NP_001101941.1 | 2 | 763 | ||
GenBank mRNA | FQ209516.1 | - | 3 | 356 |