siRNA Details
Gene Symbol: Ube3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191837.2 | NP_001178766.1 | 8 | 2429 | ||
XM_006229310.2 | XP_006229372.1 | 7 | 3274 | |||
XM_006229313.2 | XP_006229375.1 | 6 | 3154 | |||
XM_008759399.1 | XP_008757621.1 | 5 | 3984 | |||
XM_008759400.1 | XP_008757622.1 | 7 | 1915 | |||
XM_008759401.1 | XP_008757623.1 | 7 | 3218 |
