siRNA Details
Gene Symbol: Fam168a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108494.1 | NP_001101964.1 | 7 | 744 | ||
XM_006229773.2 | XP_006229835.1 | 6 | 1901 |

More Information
Other Species Targeted

