siRNA Details
Gene Symbol: Relt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108495.1 | NP_001101965.1 | 2, 3 | 161 | ||
XM_006229775.2 | XP_006229837.1 | 3, 4 | 329 | |||
XM_006229776.2 | XP_006229838.1 | 3, 4 | 2593 | |||
XM_006229777.2 | XP_006229839.1 | 3, 4 | 280 | |||
XM_006229778.2 | XP_006229840.1 | 3, 4 | 4681 | |||
XM_006229780.2 | XP_006229842.1 | 3, 4 | 329 | |||
XM_008759763.1 | XP_008757985.1 | 3, 4 | 5009 | |||
XM_008759764.1 | XP_008757986.1 | 3, 4 | 4934 | |||
XM_008759765.1 | XP_008757987.1 | 3, 4 | 4889 |
