siRNA Details
Gene Symbol: Trim66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191839.1 | NP_001178768.1 | 10 | 2149 | ||
XM_006229983.2 | XP_006230045.1 | 11 | 3346 | |||
GenBank mRNA | BC168212.1 | - | 11 | 2036 |
