siRNA Details
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047906.2 | NP_001041371.2 | 7 | 991 | ||
GenBank mRNA | AY325173.1 | - | 741 | |||
BC099199.1 | - | 7 | 965 | 7383928 | ||
FQ225106.1 | - | 7 | 971 |

More Information
Other Species Targeted

Gene Ontology Categories:
