siRNA Details
Gene Symbol: Cutc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108525.1 | NP_001101995.1 | 7, 8 | 739 | ||
XM_006231526.1 | XP_006231588.1 | 7, 8 | 728 | |||
XM_006231527.1 | XP_006231589.1 | 6, 7 | 692 |
